Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC4C Polyclonal Antibody
Rabbit Polyclonal Antibody
$597.00
Specifications
Antigen | LRRC4C |
---|---|
Concentration | 0.20 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111400
|
Invitrogen™
PA5111400 |
100 μL |
Each of 1 for $597.00
|
|
|||||
Description
Immunogen sequence: NVDDEITGDT PMESHLPMPA IEHEHLNHYN SYKSPFNHTT TVNTINSIHS SVHEPLLIRM NSKD.
Leucine-rich repeat-containing 4C (LRRC4C), also known as Netrin-G ligand 2 (NGL-2), is a gene highlighted for its expression in the nervous system, particularly the brain. It is a member of the leucine-rich repeat superfamily, which plays a significant role in cell adhesion and signaling pathways critical for synaptic connectivity and neuronal development. LRRC4C is involved in neural processes such as axon guidance and synapse formation, aiding in the establishment of functional neural circuits. Additionally, research has suggested that LRRC4C may function as a tumor suppressor, particularly in gliomas, indicating its potential role in neuro-oncological disorders. Variations or deletions in this gene have been linked to neurodevelopmental disorders, demonstrating its importance in normal brain function and development. Furthermore, LRRC4C's expression patterns and associations with cancer also suggest its potential as a prognostic marker in clinical settings.Specifications
LRRC4C | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
LRRC4C | |
6430556C10Rik; KIAA1580; leucine rich repeat containing 4C; leucine-rich repeat-containing protein 4C; Lrrc4c; Netrin-G1 ligand; NGL1; NGL-1; RGD1311013; UNQ292/PRO331 | |
LRRC4C | |
Primary | |
Antigen affinity chromatography |
0.20 mg/mL | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCJ2 | |
57689 | |
Recombinant Protein corresonding to Human LRRC4C. Recombinant protein control fragment (Product #RP-106572). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title