Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC50 Antibody (OTI2C4), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB278751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278751 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278751 Supplier Novus Biologicals Supplier No. NBP272536MFV450

Mouse Monoclonal Antibody

LRRC50 Monoclonal antibody specifically detects LRRC50 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry

Specifications

Antigen LRRC50
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2C4
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human LRRC50(NP_848547) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 123872
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.