Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC52 Antibody (1037552), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB175835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175835 Supplier Novus Biologicals Supplier No. FAB10260E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRC52 Monoclonal specifically detects LRRC52 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC52
Applications Flow Cytometry
Classification Monoclonal
Clone 1037552
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ25811, leucine rich repeat containing 52, leucine-rich repeat-containing protein 52
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human LRRC52, Ser24-Asp244, Accession # Q8N7C0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440699
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.