Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC6 Antibody (OTI7E9), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272542IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272542IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272542IR Supplier Novus Biologicals Supplier No. NBP272542IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRRC6 Monoclonal specifically detects LRRC6 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen LRRC6
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI7E9
Conjugate DyLight 755
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias leucine rich repeat containing 6, Leucine-rich testis-specific protein, LRTPleucine-rich repeat-containing protein 6, Testis-specific leucine-rich repeat protein, TSLRPtestis specific leucine rich repeat protein
Gene Symbols LRRC6
Host Species Mouse
Immunogen Full length recombinant protein of human LRRC6 (NP_036604) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23639.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.