Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC74A Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB352907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352907 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352907 Supplier Novus Biologicals Supplier No. NBP306240MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC74A Polyclonal antibody specifically detects LRRC74A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen LRRC74A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C14orf166B, chromosome 14 open reading frame 166B, hypothetical protein LOC145497, leucine rich repeat containing 74, leucine-rich repeat-containing protein 74, leucine-rich repeat-containing protein 74A, LRRC74
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human LRRC74A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145497
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.