Learn More
Description
Specifications
Specifications
| Antigen | LRRC8A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8 |
| Gene Symbols | LRRC8A |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 (GeneID 56262). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
