Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC8B Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB326470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326470 Supplier Novus Biologicals Supplier No. NBP299221PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC8B Polyclonal antibody specifically detects LRRC8B in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen LRRC8B
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRRC8B.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23507
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.