Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRFIP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP171835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171835 Supplier Novus Biologicals Supplier No. NBP171835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRFIP1 Polyclonal specifically detects LRRFIP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen LRRFIP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence -From verified customer review., Immunoprecipitation 2 - 5 ug/mg lysate
Formulation TBS, 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_001131024.1
Gene Alias FLAP-1, FLIIAP1, GC-binding factor 2, GCF2, GCF-2, HUFI-1, leucine rich repeat (in FLII) interacting protein 1, leucine-rich repeat flightless-interacting protein 1, LRR FLII-interacting protein 1, MGC10947, MGC119738, MGC119739, NEDD8-conjugating enzyme, TRIPTAR RNA-interacting protein
Gene Symbols LRRFIP1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 500 and 550 of human Leucine Rich Repeat (in FLII) Interacting Protein 1 using the numbering given in entry NP_001131024.1 (GeneID 9208).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9208
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.