Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRFIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166128 20 μg
NB166127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166128 Supplier Novus Biologicals Supplier No. NBP31551620UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRFIP2 Polyclonal antibody specifically detects LRRFIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen LRRFIP2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 50% glycerol, pH7.3
Gene Alias DKFZp434H2035, FLJ20248, FLJ22683, FLJ58304, HUFI-2, leucine rich repeat (in FLII) interacting protein 2, leucine-rich repeat flightless-interacting protein 2, LRR FLII-interacting protein 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-340 of human LRRFIP2 (NP_060194.1). VIEEQEEQMAEFYRENEEKSKELERQKHMCSVLQHKMEELKEGLRQRDELIEKHGLVIIPDGTPNGDVSHEPVAGAITVVSQEAAQVLESAGEGPLDVRLRKLAGEKEELLSQIRKLKLQLEEERQKCSRNDGTVGDLAGLQNGSDLQFIEMQRDANRQISEYKFKLSKAEQDITTLEQSISRLEGQVLRY
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9209
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.