Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB11058771T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058771T 0.025 mL
NB11058771 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11058771T Supplier Novus Biologicals Supplier No. NB11058771SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, Knockdown Validated.

Specifications

Antigen LRRK2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunoassay
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24312256), Immunohistochemistry-Frozen, Flow (Cell Surface) reported in scientific literature (PMID 20483355), Knockout Validated, KnockDown Validated
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.05% Sodium Azide
Gene Accession No. Q5S007
Gene Alias augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2
Gene Symbols LRRK2
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region, within residues 2500-2527 of the human LRRK2 protein, conjugated to KLH. [Swiss-Prot# Q5S007]
Molecular Weight of Antigen 286 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 120892
Test Specificity Human, rat, mouse and primate. Predicted to react with porcine based on 100% sequence homology.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.