Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB11055289W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11055289W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11055289W Supplier Novus Biologicals Supplier No. NB11055289AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRK2 Polyclonal specifically detects LRRK2 in Mouse, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LRRK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2
Gene Symbols LRRK2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the mouse LRRK2 protein sequence (between residues 900-1000). [Swiss-Prot: Q5S006]
Molecular Weight of Antigen 285 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 120892
Target Species Mouse, Human (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.