Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRK2, p Ser935 Antibody (SR1453), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32151950UL
Description
LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
LRRK2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
SR1453 | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
A synthesized peptide derived from human phospho-LRRK2 [Ser935] (Uniprot #: Q5S007) | |
50 μg | |
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
120892 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction