Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRK2, p Ser935 Antibody (SR1453), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
| Antigen | LRRK2 |
|---|---|
| Clone | SR1453 |
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
Description
LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| LRRK2 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| A synthesized peptide derived from human phospho-LRRK2 [Ser935] (Uniprot #: Q5S007) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1453 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 120892 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title