Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRN4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093382 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093382 Supplier Novus Biologicals Supplier No. NB10093382
Add to Cart
Add to Cart

Goat Polyclonal Antibody

LRRN4 Polyclonal specifically detects LRRN4 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen LRRN4
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_689824.1
Gene Alias C20orf75, chromosome 20 open reading frame 75, dJ1056H1.1, FLJ23994, leucine rich repeat neuronal 4, leucine-rich repeat neuronal protein 4, MGC25027, Neuronal leucine-rich repeat protein 4, NLRR4, NLRR-4
Gene Symbols LRRN4
Host Species Goat
Immunogen Peptide with sequence C-YKNPAFDDYP corresponding to C-Terminus according to NP_689824.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164312
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.