Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRTM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB301488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301488 Supplier Novus Biologicals Supplier No. NBP324964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRTM1 Polyclonal antibody specifically detects LRRTM1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LRRTM1
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias FLJ32082, leucine rich repeat transmembrane neuronal 1, leucine-rich repeat transmembrane neuronal 1 protein, leucine-rich repeat transmembrane neuronal protein 1
Host Species Rabbit
Immunogen This antibody has been engineered to specifically recognize the recombinant protein LRRTM1 using the following amino acid sequence: EPHVFETVPHLQSLQLDSNRLTYIEPRILNSWKSLTSITLAGNLWDCGRNVCALASWLNNFQGRYDGNLQCASPEYAQGEDVLDAVYAFHLCEDGAEPTSGHLLSAVTNRSDLGPPASSATTLADGGEGQHDGT
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 347730
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.