Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRTM1 (extracellular) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5111862
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111862 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111862 Supplier Thermo Scientific Supplier No. PA5111862

Rabbit Polyclonal Antibody

Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)GRYDANL Q(S)ASPE Homology: Mouse and rat - 12/13 amino acid residues identical; human - 11/13 amino acid residues identical.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM1 is a maternally suppressed gene that is associated paternally with handedness and schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM1 (extracellular)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.05% sodium azide; pH 7.4
Gene LRRTM1
Gene Accession No. D4A6D8, Q8K377
Gene Alias 4632401D06Rik; AW125451; Leucine-rich repeat transmembrane neuronal protein 1; LRRTM1; UNQ675/PRO1309
Gene Symbols LRRTM1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid residues 334-346 of mouse LRRTM1 (Extracellular, N-terminus)
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 679668, 74342
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.