Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRTM1 Antibody (2381D) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB402587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402587 Supplier Novus Biologicals Supplier No. FAB10046A
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LRTM1 Monoclonal antibody specifically detects LRTM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen LRTM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 2381D
Conjugate APC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1
Host Species Rabbit
Immunogen Synthetic peptide containing human LRTM1, Accession # Q9HBL6
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57408
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.