Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSD1 Antibody (1B2E5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
Supplier: Novus Biologicals NB1001762SS
Description
LSD1 Monoclonal specifically detects LSD1 in Human, Mouse, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LSD1 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
Mouse | |
93 kDa | |
0.025 mL | |
Chromatin Modifiers, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
23028 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay | |
1B2E5 | |
Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP), KnockDown Validated | |
O60341 | |
KDM1A | |
Purified recombinant fragment of human LSD1 (between amino acids 400-600) expressed in E. coli. [UniProt# O60341] | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse, Primate | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction