Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSD1 Antibody (1B2E5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 11 publications

$206.50 - $499.50

Specifications

Antigen LSD1
Clone 1B2E5
Dilution Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP), KnockDown Validated
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001762SS
SDP
View Documents
Novus Biologicals
NB1001762SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB1001762
SDP
View Documents
Novus Biologicals
NB1001762
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

LSD1 Monoclonal specifically detects LSD1 in Human, Mouse, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

LSD1
Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP), KnockDown Validated
Monoclonal
Ascites
RUO
Human, Mouse, Primate
O60341
23028
Purified recombinant fragment of human LSD1 (between amino acids 400-600) expressed in E. coli. [UniProt# O60341]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1B2E5
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Unconjugated
Mouse
Chromatin Modifiers, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Ascites with 0.03% Sodium Azide
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A
KDM1A
IgG1
Unpurified
93 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.