Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSECtin/CLEC4G Antibody (845404), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB363611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363611 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363611 Supplier Novus Biologicals Supplier No. FAB2947Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSECtin/CLEC4G Monoclonal specifically detects LSECtin/CLEC4G in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LSECtin/CLEC4G
Applications Flow Cytometry
Classification Monoclonal
Clone 845404
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, UNQ431
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G, Ser54-Cys293, Accession # Q6UXB4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339390
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.