Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM1 Antibody (OTI5C6), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272547PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272547PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272547PEC5 Supplier Novus Biologicals Supplier No. NBP272547PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSM1 Monoclonal specifically detects LSM1 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen LSM1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5C6
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cancer-associated Sm-like, CASMYJL124C, LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae), Small nuclear ribonuclear CaSm, U6 snRNA-associated Sm-like protein LSm1
Gene Symbols LSM1
Host Species Mouse
Immunogen Full length recombinant protein of human LSM1 (NP_055277) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 27257.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.