Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM10 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095931 100 μL
NB095932 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095931 Supplier Novus Biologicals Supplier No. NBP304424100UL

Rabbit Polyclonal Antibody

LSM10 Polyclonal specifically detects LSM10 in Rat samples. It is validated for Western Blot.

Specifications

Antigen LSM10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias LSM10, U7 small nuclear RNA associated, MGC15749, MST074, MSTP074, U7 snRNA-associated Sm-like protein LSm10, U7 snRNP-specific Sm-like protein LSM10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human LSM10 (NP_116270.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84967
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.