Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM14A Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB326854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326854 Supplier Novus Biologicals Supplier No. NBP299157APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSM14A Polyclonal antibody specifically detects LSM14A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen LSM14A
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias alphaSNBP, C19orf13, DKFZp434D1335, FAM61A, family with sequence similarity 61, member A, hRAP55, hRAP55A, LSM14 homolog A, LSM14A, SCD6 homolog A (S. cerevisiae), Protein FAM61A, protein LSM14 homolog A, Protein SCD6 homolog, Putative alpha-synuclein-binding protein, RAP55ALSM14 homolog A (SCD6, S. cerevisiae), RAP55chromosome 19 open reading frame 13, RNA-associated protein 55, RNA-associated protein 55A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LSM14A.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26065
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.