Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226136 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226136 Supplier Novus Biologicals Supplier No. NBP226136
Add to Cart
Add to Cart

Goat Polyclonal Antibody

LSM2 Polyclonal specifically detects LSM2 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen LSM2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias C6orf28chromosome 6 open reading frame 28, G7b, LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae), Protein G7b, Small nuclear ribonuclear protein D homolog, snRNP, snRNP core Sm-like protein Sm-x5, U6 snRNA-associated Sm-like protein LSm2, YBL026W
Gene Symbols LSM2
Host Species Goat
Immunogen Peptide with sequence C-DQYLNIKLTDISVT corresponding to internal region according to NP_067000.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57819
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.