Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB092059 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092059 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB092059 Supplier Novus Biologicals Supplier No. NBP300316AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSM3 Polyclonal specifically detects LSM3 in Human samples. It is validated for ELISA.

Specifications

Antigen LSM3
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae), SMX4, U6 snRNA-associated Sm-like protein LSm3, USS2, YLR438C
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant LSM3 (P62310; Met1-Gly102).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 27258
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.