Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM5 Antibody (25F8), PerCP, Novus Biologicals™
SDP

Catalog No. NB309056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309056 Supplier Novus Biologicals Supplier No. NBP259399PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSM5 Monoclonal antibody specifically detects LSM5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LSM5
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 25F8
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias FLJ12710, LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae), U6 snRNA-associated Sm-like protein LSm5, YER146W
Host Species Mouse
Immunogen Recombinant human LSM5 (1-91aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23658
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.