Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSMD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174258
Description
LSMD1 Polyclonal specifically detects LSMD1 in Human samples. It is validated for Western Blot.Specifications
| LSMD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| LSM domain containing 1, MAK31, MGC14151, MGC74837, PFAAP2LSM domain-containing protein 1, Phosphonoformate immuno-associated protein 2, regulated by phosphonoformate | |
| Rabbit | |
| 19 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: ; Dog: 86%; Horse: 86%; Rabbit: 86%; Bovine: 80%; Pig: 79%; Rat: 79%; Mouse: 79%; Guinea pig: 79%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BRA0-2 | |
| LSMD1 | |
| Synthetic peptides corresponding to the middle region of LSMD1. Immunizing peptide sequence RAAWAARSERLLHPAQDSDGEREDSAAERARQQLEALLNKTMRIRMTDGR. | |
| Affinity purified | |
| RUO | |
| 84316 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction