Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSMD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | LSMD1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LSMD1 Polyclonal specifically detects LSMD1 in Human samples. It is validated for Western Blot.Specifications
| LSMD1 | |
| Polyclonal | |
| Rabbit | |
| Q9BRA0-2 | |
| 84316 | |
| Synthetic peptides corresponding to the middle region of LSMD1. Immunizing peptide sequence RAAWAARSERLLHPAQDSDGEREDSAAERARQQLEALLNKTMRIRMTDGR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| LSM domain containing 1, MAK31, MGC14151, MGC74837, PFAAP2LSM domain-containing protein 1, Phosphonoformate immuno-associated protein 2, regulated by phosphonoformate | |
| LSMD1 | |
| IgG | |
| 19 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title