Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP174048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174048 0.1 mg
NBP174048SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP174048 Supplier Novus Biologicals Supplier No. NBP174048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSP1 Polyclonal specifically detects LSP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen LSP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 - 2.0 ug/ml, ELISA 0.01-0.1 ug/ml, Immunohistochemistry 1:200, Immunoprecipitation 2-5 ug/ml, Immunohistochemistry-Paraffin 1:200
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P33241
Gene Alias 47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34
Gene Symbols LSP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding a C-terminal portion of the human LSP1 protein (between amino acids 250-339) [UniProt P33241].
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4046
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.