Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LST1 Antibody (LST1/02), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP145072Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145072Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145072Z Supplier Novus Biologicals Supplier No. NBP145072AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LST1 Monoclonal specifically detects LST1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LST1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone LST1/02
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias B144D6S49ELST-1, leucocyte specific transcript 1, leukocyte specific transcript 1, leukocyte-specific transcript 1 protein, lymphocyte antigen 117, MGC119006, MGC119007, Protein B144
Gene Symbols LST1
Host Species Mouse
Immunogen KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7940
Test Specificity The antibody LST1/02 reacts with an extracellular epitope of LST1, an approximately 6-11 kDa protein expressed as various transmembrane or soluble forms. LST1 is found predominantly on monocytes and dendritic cells. It migrates on SDS PAGE gels as approximately 25-28 kDa molecule.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.