Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lubricin/PRG4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074453 Supplier Novus Biologicals Supplier No. NB10074453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lubricin/PRG4 Polyclonal specifically detects Lubricin/PRG4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Lubricin/PRG4
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS, 1mg/ml BSA with 0.05% Sodium Azide
Gene Alias bG174L6.2, bG174L6.2 (MSF: megakaryocyte stimulating factor ), CACP, FLJ32635, JCAP, lubricin, megakaryocyte stimulating factor, Megakaryocyte-stimulating factor, MSFproteoglycan 4, (megakaryocyte stimulating factor, articular superficial zoneprotein, camptodactyly, arthropathy, coxa vara, pericarditis syndrome), proteoglycan 4, Superficial zone proteoglycan, SZPHAPOarticular superficial zone protein
Gene Symbols PRG4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C L(1356) P N I R K Q P D G Y D Y Y A F S K D Q(1374) of human Lubricin.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10216
Test Specificity Detects Lubricin in human samples.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.