Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lubricin/PRG4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP119048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119048 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119048 Supplier Novus Biologicals Supplier No. NBP119048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Lubricin/PRG4 Polyclonal specifically detects Lubricin/PRG4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Lubricin/PRG4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation Antiserum with No Preservative
Gene Accession No. Q92954
Gene Alias bG174L6.2, bG174L6.2 (MSF: megakaryocyte stimulating factor ), CACP, FLJ32635, JCAP, lubricin, megakaryocyte stimulating factor, Megakaryocyte-stimulating factor, MSFproteoglycan 4, (megakaryocyte stimulating factor, articular superficial zoneprotein, camptodactyly, arthropathy, coxa vara, pericarditis syndrome), proteoglycan 4, Superficial zone proteoglycan, SZPHAPOarticular superficial zone protein
Gene Symbols PRG4
Host Species Rabbit
Immunogen Synthetic peptide derived from N terminal domain of PRG4 protein
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10216
Test Specificity Reacts specifically with human 151 kDa protein
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.