Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB12063410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12063410 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12063410 Supplier Novus Biologicals Supplier No. NB1206341.0MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Luciferase Polyclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Luciferase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:8000-1:31000, ELISA 1:80000-1:310000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Goat
Immunogen Native protein from Photinus pyralis (Firefly).
Purification Method Delipidation and Defibrination
Quantity 1 mg
Research Discipline Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Test Specificity Luciferase, no reactivity is observed against Sea pansy (Renilla reniformis) luciferase.
Target Species Firefly
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.