Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB100167601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100167601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100167601 Supplier Novus Biologicals Supplier No. NB10016760.1MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Luciferase Polyclonal specifically detects Luciferase in Firefly samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Luciferase
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot 1:5000-1:10000, ELISA 1:50000-1:100000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Goat
Immunogen Native protein from Photinus pyralis (Firefly).
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Test Specificity Luciferase, no reactivity is observed against Sea pansy (Renilla reniformis) luciferase.
Target Species Firefly
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.