Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase Antibody (Luci 21 1-107), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB600307AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600307AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600307AI Supplier Novus Biologicals Supplier No. NB600307AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Luciferase Monoclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Luciferase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Luci 21 1-107
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Mouse
Immunogen This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659].
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Test Specificity This Luciferase Antibody (Luci 21 1-107) is specific for Luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time.
Target Species Firefly
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.