Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase Antibody, Novus Biologicals™
SDP

Catalog No. NBP24416505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24416505 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24416505 Supplier Novus Biologicals Supplier No. NBP2441650.5ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Luciferase Polyclonal specifically detects Luciferase in Bacteria samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot.

Specifications

Antigen Luciferase
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, ELISA 1:5000 - 1:25000, Immunohistochemistry 1:500 - 1:2500, Immunoprecipitation 1:100, Dot Blot
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Rabbit
Immunogen Luciferase [Photobacterium fischerii]. Uniprot ID P19907
Quantity 0.5 mL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.