Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Luciferase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$402.00
Specifications
Antigen | Luciferase |
---|---|
Dilution | Western Blot 1:1000 - 1:5000, ELISA 1:5000 - 1:25000, Immunohistochemistry 1:500 - 1:2500, Immunoprecipitation 1:100, Dot Blot |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24416505
![]() |
Novus Biologicals
NBP2441650.5ML |
0.5 mL |
Each of 1 for $402.00
|
|
|||||
Description
Luciferase Polyclonal specifically detects Luciferase in Bacteria samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot.Specifications
Luciferase | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot | |
Unconjugated | |
Bacteria | |
ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
Primary |
Western Blot 1:1000 - 1:5000, ELISA 1:5000 - 1:25000, Immunohistochemistry 1:500 - 1:2500, Immunoprecipitation 1:100, Dot Blot | |
Polyclonal | |
Rabbit | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
Luciferase [Photobacterium fischerii]. Uniprot ID P19907 | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title