Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Luciferin Detection Reagent

Catalog No. PRV8920 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8920 10 mL
PRV8921 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8920 Supplier Promega Supplier No. V8920
Add to Cart
Add to Cart

A reagent to detect presence of pro-luciferin substrates used to monitor the activity of specific isoforms of cytochrome P450 or NAT2.

  • Reagent for Detection of Pro-Luciferin Substrates
  • Reagent available without esterase
  • Use in combination with the pro-luciferin substrates

Specifications

Quantity 10 mL
Content And Storage -30°C to -10°C
For Use With (Application) Monitor the activity of specific isoforms of cytochrome P450 or NAT2
Kit Contents 10mL luciferin detection reagent and 10mL reconstitution buffer
pH 7.2
Product Type Luciferin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.