Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris Color Probe qPCR Master Mix

Catalog No. FERK0354
Change view
Click to view available options
No. of Reactions:
1,250 Reactions
250 Reactions
5,000 Reactions
500 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
FERK0354 5,000 Reactions
FERK0353 1,250 Reactions
FERK0351 250 Reactions
FERK0352 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK0354 Supplier Thermo Scientific™ Supplier No. K0354

Optimize qPCR with probe detection using standard cycling protocols with these ready-to-use master mixes with premixed UDG, available with or without blue color.

Luminaris Color Probe and Luminaris Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.

The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.

The use of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.

Thermo Scientific Luminaris Color Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.

The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.

The use of Luminaris Color Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.

  • Specificity no primer dimers

  • Sensitivitity detects low copy number targets

  • Wide linear range accurate quantification across 9 orders of magnitude

  • Reproducibility and convenience ready-to-use 2X master mix

  • Blue Master Mix and Yellow Sample Buffer for easy pipetting
  • UDG in the master mix to prevent carry-over contamination
  • Excellent qPCR performance
  • Versions with premixed ROX for different real-time platforms
  • Standard cycling protocols
Luminaris Color Probe High ROX qPCR Master Mix (2X) and Luminaris Color Probe Low ROX qPCR Master Mix (2X) contain different amounts of ROX passive reference dye for specific real-time platforms.

Includes:

  • Luminaris Color Probe qPCR Master Mixes include Hot Start Taq Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer with a blue dye
  • 40X Yellow Sample Buffer with a yellow dye
  • Nuclease-free water

Recommended for:

Gene expression analysis; siRNA validation; Genotyping; Pathogen detection

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X (qPCR Master Mix), 40X (Buffer)
Content And Storage Contains:
• 4 x 12.5 mL Luminaris Color Probe qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG and dNTPs (with dUTP) in an optimized PCR buffer with a blue dye; sufficient for 5000 x 20 μL reactions
• 1 x 13 mL 40X Yellow Sample Buffer
• 2 x 30 mL Nuclease-Free Water

Store at -20°C.
Description Color Probe
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product Line Luminaris
Product Type qPCR Master Mix
Quantity 5000 x 20 μL Reactions
Sufficient For 5000 Reactions of 20 μL
For Use With (Application) Gene Expression
No. of Reactions 5,000 Reactions
Sample Type DNA, dsDNA
Volume 4 x 12.5 mL
Show More Show Less
Does dUTP in Luminaris qPCR master mix influence qPCR efficiency?

Luminaris qPCR master mixes have all dTTP substituted with dUTP. dUTP presence does not influence the reaction speed and efficiency of the master mix. 100% substitution of dTTP with dUTP facilitates an efficient clearance of the contamination from previous PCR reactions.

What enzyme is used in Luminaris qPCR Master Mixes?

Luminaris qPCR master mixes contain Hot Start Taq DNA Polymerase. This is a Taq DNA polymerase, which has been chemically modified by the addition of heat-labile groups to amino acid residues blocking enzyme activity. The Hot Start Taq DNA Polymerase has ultra low residual activity until thermally activated. Therefore reactions can be assembled at room temperature with no risk for the extension of non-specifically annealed primers or primer dimers and providing higher specificity for DNA amplification. The enzyme is activated by 10 minutes incubation at 95 degrees C.

Can I prepare a qPCR plate in advance and store it for later use when using Luminaris qPCR Master Mixes?

When using a hot start polymerase it is not critical to start reactions immediately after setup. The Hot Start Taq DNA Polymerase in Luminaris qPCR Master Mixes has ultra low residual activity until thermally activated. Therefore reactions can be assembled and stored at room temperature for up to 72 hours with no risk for the extension of non-specifically annealed primers or primer dimers.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.