Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Luminogenic Enzyme Substrates
Shop All Promega Corporation Products
Click to view available options
Substrate Quantity:
3mg
Substrate Type:
Luciferin-2J2/4F12 (ester)
Luciferin-3A7 Substrate
Luciferin-4A Substrate
Luciferin-4F12 Substrate
Luciferin-4F2/3 Substrate
Description
- Pro-luciferin Substrates to Monitor Activity of Cytochrome P450 Isoforms
- Used to monitor the activity of specific isoforms of cytochrome P450 or NAT2
- Choose from a wide range of substrates
- Cat.# P1621 is discontinued.
Specifications
Specifications
| For Use With (Application) | Monitor the activity of specific isoforms of cytochrome P450 or NAT2 |
| Quantity | 3mg |
| Substrate Type | Luciferin-2J2/4F12 (ester) |
| Storage Requirements | −30 to −10°C |
| Substrate Quantity | 3mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction