Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lunatic Fringe Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Lunatic Fringe |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15943220
![]() |
Novus Biologicals
NBP15943220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159432
![]() |
Novus Biologicals
NBP159432 |
100 μL |
Each for $487.50
|
|
|||||
Description
Lunatic Fringe Polyclonal specifically detects Lunatic Fringe in Human samples. It is validated for Western Blot.Specifications
Lunatic Fringe | |
Polyclonal | |
Rabbit | |
Q8NES3-3 | |
3955 | |
Synthetic peptides corresponding to LFNG(LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase) The peptide sequence was selected from the N terminal of LFNG. Peptide sequence LSEYFSLLTRARRDAGPPPGAAPRPADGHPRPLAEPLAPRDVFIAVKTTK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
beta-1,3-N-acetylglucosaminyltransferase lunatic fringe, LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, lunatic fringe (Drosophila) homolog, lunatic fringe homolog (Drosophila), O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, SCDO3EC 2.4.1.222 | |
LFNG | |
IgG | |
39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title