Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUZP4 Antibody (OTI6D11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272549G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272549G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272549G Supplier Novus Biologicals Supplier No. NBP272549G

Mouse Monoclonal Antibody

LUZP4 Monoclonal specifically detects LUZP4 in Human samples. It is validated for Western Blot.

Specifications

Antigen LUZP4
Applications Western Blot
Classification Monoclonal
Clone OTI6D11
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias leucine zipper protein 4
Gene Symbols LUZP4
Host Species Mouse
Immunogen Full length recombinant protein of human LUZP4 (NP_057467) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51213.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.