Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR alpha/NR1H3 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB400157C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400157C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400157C Supplier Novus Biologicals Supplier No. NB400157C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LXR alpha/NR1H3 Polyclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot.

Specifications

Antigen LXR
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q13133
Gene Alias alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1
Gene Symbols NR1H3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt# Q13133]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10062
Test Specificity Human, mouse, porcine and monkey. Immunogen sequence has 92% identity to rat and 90% identity to chicken.
Target Species Human, Mouse, Pig, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.