Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR beta/NR1H2 Antibody (LXRB/2731), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB378313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378313 Supplier Novus Biologicals Supplier No. NBP308914IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LXR beta/NR1H2 Monoclonal antibody specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen LXR beta/NR1H2
Applications ELISA, Protein Array
Classification Monoclonal
Clone LXRB/2731
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta
Host Species Mouse
Immunogen Recombinant full-length human LXR beta/NR1H2 protein (Uniprot: P55055)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7376
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.