Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR beta/NR1H2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074457 Supplier Novus Biologicals Supplier No. NB10074457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

LXR beta/NR1H2 Polyclonal specifically detects LXR beta/NR1H2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen LXR beta/NR1H2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Formulation PBS with 1mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P55055
Gene Alias Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta
Gene Symbols NR1H2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues R(113) Y A C R G S G T C Q M D A(126) of rat LXR beta.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7376
Test Specificity Detects LXR beta in rat and mouse samples.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.