Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | Ly-108 |
---|---|
Clone | eBio13G3-19D (13G3-19D) |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M050T03Y06A
![]() |
Invitrogen™
M050T03Y06A |
100 μg |
Each for $379.00
|
|
|||||
M050T02Y06A
![]() |
Invitrogen™
M050T02Y06A |
25 μg |
Each for $201.00
|
|
|||||
Description
Ly-108 Monoclonal antibody specifically detects Ly-108 in Mouse samples. It is validated for Flow CytometrySpecifications
Ly-108 | |
0.1 mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
Q9ET39 | |
30925 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBio13G3-19D (13G3-19D) | |
Flow Cytometry | |
NovaFluor Yellow 700 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
IgG2a κ | |
Affinity chromatography | |
SLAMF6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title