Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LY 2033298

Click to view available options
Quantity:
5 mg
25 mg
Description
LY 2033298 is a selective positive allosteric modulator of M4 receptor (KB = 200 nM, α = 35); increases the potency of acetylcholine by 40-fold. Exhibits no effect at hM1/3/5 receptors and a small effect at hM2 (KB = 1 μM α = 3.7). Potentiates other full and partial orthosteric agonists, including Carbachol, Oxotremorine-M, and McN-A343 . Does not affect binding of antagonist NMS. Active in vivo. Antipsychotic.
α indicates the degree of allosteric enhancement when both orthosteric and allosteric sites are occupied (derived from the allosteric ternary complex model).
Specifications
Specifications
| Chemical Name or Material | 3-Amino-5-chloro-N-cyclopropyl-6-methoxy-4-methylthieno[2,3-b]pyridine-2-carboxamide |
| CAS | 886047-13-8 |
| Quantity | 5 mg |
| Molecular Formula | C13H14ClN3O2S |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction