Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 2365109 hydrochloride
GREENER_CHOICE

Catalog No. 339710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-971-0 10 mg
33-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-971-0 Supplier Tocris Bioscience™ Supplier No. 3397/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective GlyT1 inhibitor

Potent and selective glycine transporter 1 (GlyT1) inhibitor (IC50 values are 15.8 and > 30 000 nM at GlyT1 and GlyT2 respectively). Induces a dose-dependent elevation in CSF levels of glycine, and enhances acetylcholine and dopamine release in the striatum and prefrontal cortex respectively. Produces profound locomotor and respiratory impairments at higher doses.

Chemical Identifiers

CAS 868265-28-5
Molecular Formula C22H28ClNO5

Specifications

Quantity 10 mg
Formula Weight 421.91
Purity >98%
Molecular Weight (g/mol) 421.918
Chemical Name or Material LY 2365109 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.