Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 266097 hydrochloride
GREENER_CHOICE

Catalog No. 408110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-811-0 10 mg
40-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-811-0 Supplier Tocris Bioscience™ Supplier No. 4081/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective 5-HT2B receptor antagonist

Selective 5-HT2B receptor antagonist (pKi = 9.3). More than 100-fold selective over 5-HT2A and 5-HT2C. Attenuates amphetamine-induced locomotion in the rat.

Chemical Identifiers

CAS 172895-39-5
Molecular Formula C21H24Cl2N2O2

Specifications

Quantity 10 mg
Formula Weight 407.33
Purity >99%
Molecular Weight (g/mol) 407.335
Chemical Name or Material LY 266097 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.