Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 288513
GREENER_CHOICE

Catalog No. 152410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-241-0 10 mg
15-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-241-0 Supplier Tocris Bioscience™ Supplier No. 1524/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective CCK2 antagonist

Selective CCK2 receptor antagonist (IC50 values are 16 and > 30,000 nM for CCK2 and CCK1 respectively). Displays anxiolytic and antipsychotic properties in vivo.

Chemical Identifiers

CAS 147523-65-7
Molecular Formula C22H18BrN3O2

Specifications

Quantity 10 mg
Formula Weight 436.3
Purity >98%
Molecular Weight (g/mol) 436.309
Chemical Name or Material LY 288513
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.